Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021265.3 | NP_067088.2 | 7 | 642 | ||
XM_006245011.2 | XP_006245073.1 | 8 | 1929 | |||
XM_008767130.1 | XP_008765352.1 | 8 | 1929 | |||
XM_008767131.1 | XP_008765353.1 | 9 | 1069 | |||
XM_008767132.1 | XP_008765354.1 | 10 | 955 | |||
XM_008767133.1 | XP_008765355.1 | 6 | 524 | |||
GenBank mRNA | AB030499.1 | - | 5 | 591 | ||
AB030500.1 | - | 5 | 591 | |||
AB030501.1 | - | 5 | 591 |