Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIN3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297595.1 | NP_001284524.1 | 13 | 2250 | ||
NM_001297597.1 | NP_001284526.1 | 4 | 1356 | |||
NM_015260.3 | NP_056075.1 | 14 | 2346 | |||
XM_006722704.1 | XP_006722767.1 | 4 | 961 | |||
GenBank mRNA | AB014600.2 | - | 13 | 2237 | ||
AK001633.1 | - | 7 | 1351 | |||
AK295618.1 | - | 4 | 1295 | |||
AY706204.1 | - | 14 | 2326 | |||
BC005113.1 | - | 1 | 88 |