Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC360919 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751340.3 | XP_003751388.1 | 13, 14 | 1987 | ||
XM_006250759.2 | XP_006250821.1 | 13, 14 | 1985 | |||
XM_006250761.2 | XP_006250823.1 | 11, 12 | 1765 |