Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126094.2 | NP_001119566.1 | 2 | 411 | ||
NM_001287116.1 | NP_001274045.1 | 3 | 273 | |||
XM_006232983.2 | XP_006233045.1 | 3 | 402 | |||
GenBank mRNA | BC166891.1 | - | 3 | 273 | 6918483 | |
CB808996.1 | - | 2 | 173 | |||
FQ210042.1 | - | 3 | 249 | |||
FQ221433.1 | - | 3 | 246 | |||
FQ224554.1 | - | 3 | 249 |