Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015276.1 | NP_056091.1 | 4 | 661 | ||
XM_005256575.2 | XP_005256632.1 | 4 | 375 | |||
GenBank mRNA | AB028986.1 | - | 4 | 661 | ||
BC110499.1 | - | 4 | 420 | |||
BC126898.1 | - | 4 | 421 | 40027759 | ||
BX640815.1 | - | 3 | 243 |