Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164543.1 | NP_001158015.1 | 2 | 754 | ||
NM_001166165.1 | NP_001159637.1 | 3 | 1041 | |||
GenBank mRNA | BC158725.1 | - | 2 | 754 | 9090994 | |
FQ220528.1 | - | 2 | 518 |
Gene Symbol: LOC688724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764897.1 | XP_008763119.1 | 2 | 505 |