Search Thermo Fisher Scientific
Gene Symbol: CAMK2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015981.3 | NP_057065.2 | 3, 4 | 435 | ||
NM_171825.2 | NP_741960.1 | 3, 4 | 435 | |||
XM_017009898.1 | XP_016865387.1 | 3, 4 | 435 | |||
GenBank mRNA | AB023185.1 | - | 3, 4 | 368 | ||
AF091486.1 | - | 3, 4 | 324 | |||
AF145710.1 | - | 3, 4 | 226 | |||
AF145711.1 | - | 3, 4 | 226 | |||
AK289776.1 | - | 3, 4 | 435 | |||
BC040457.1 | - | 3, 4 | 350 | 4811948 |