Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177933.2 | NP_808794.2 | 16, 17 | 1720 | ||
GenBank mRNA | AF304853.1 | - | 17, 18 | 1734 | ||
CO562235.1 | - | 7, 8 | 609 |