Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC686031 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109490.1 | NP_001102960.1 | 1 | 364 |
Gene Symbol: LOC686042 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250422.2 | XP_006250484.1 | 1 | 399 |