Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: VPS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301138.1 | NP_001288067.1 | 12 | 1279 | ||
NM_015289.3 | NP_056104.2 | 11 | 1246 | |||
XM_011521403.2 | XP_011519705.1 | 12 | 1248 | |||
XM_011521404.2 | XP_011519706.1 | 11 | 1251 | |||
XM_017022036.1 | XP_016877525.1 | 12 | 1247 | |||
XM_017022037.1 | XP_016877526.1 | 11 | 1251 | |||
XM_017022038.1 | XP_016877527.1 | 11 | 1434 | |||
XM_017022039.1 | XP_016877528.1 | 11 | 1219 | |||
GenBank mRNA | AB018313.2 | - | 11 | 1197 | ||
AF280814.1 | - | 12 | 1106 | |||
AF281052.1 | - | 11 | 1216 | |||
BC015817.2 | - | 7 | 806 | |||
BC068559.1 | - | 11 | 1215 | 5295166 | ||
BX537404.1 | - | 11 | 1225 |