- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: VPS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301138.1 | NP_001288067.1 | 7 | 636 | ||
NM_015289.3 | NP_056104.2 | 6 | 603 | |||
XM_011521403.2 | XP_011519705.1 | 7 | 605 | |||
XM_011521404.2 | XP_011519706.1 | 6 | 608 | |||
XM_017022036.1 | XP_016877525.1 | 7 | 604 | |||
XM_017022037.1 | XP_016877526.1 | 6 | 608 | |||
XM_017022038.1 | XP_016877527.1 | 6 | 791 | |||
XM_017022039.1 | XP_016877528.1 | 6 | 576 | |||
GenBank mRNA | AB018313.2 | - | 6 | 554 | ||
AF280814.1 | - | 7 | 463 | |||
AF281052.1 | - | 6 | 573 | |||
BC015817.2 | - | 2 | 163 | |||
BC068559.1 | - | 6 | 572 | 5295166 | ||
BX537404.1 | - | 6 | 582 |