Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE1 SIRNA Assays ›
Gene Symbol:
SYNE1
Gene Name:
spectrin repeat containing nuclear envelope protein 1
Gene Aliases:
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location:
Chr.6: 152121684 - 152637399 on Build GRCh38
UniGene ID:
Hs.12967
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23609
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033071.3
NP_149062.1
130
23974
NM_182961.3
NP_892006.3
131
24352
XM_006715407.1
XP_006715470.1
131
24265
XM_006715408.2
XP_006715471.1
131
24257
XM_006715409.1
XP_006715472.1
130
24244
XM_006715410.2
XP_006715473.1
131
24269
XM_006715411.1
XP_006715474.1
130
24214
XM_006715412.2
XP_006715475.1
131
24254
XM_006715413.2
XP_006715476.1
131
24269
XM_006715414.1
XP_006715477.1
129
24193
XM_006715415.2
XP_006715478.1
131
24269
XM_006715416.2
XP_006715479.1
131
24254
XM_006715417.2
XP_006715480.1
130
24128
XM_006715420.2
XP_006715483.1
130
24116
XM_006715421.2
XP_006715484.1
130
24113
XM_006715422.1
XP_006715485.1
130
24106
XM_006715423.2
XP_006715486.1
131
24269
XM_006715424.2
XP_006715487.1
131
24269
XM_006715425.2
XP_006715488.1
131
24269
XM_011535641.2
XP_011533943.1
131
24266
XM_011535642.2
XP_011533944.1
131
24254
XM_011535643.1
XP_011533945.1
130
24100
XM_011535644.1
XP_011533946.1
117
22316
XM_011535645.2
XP_011533947.1
113
21709
XM_017010608.1
XP_016866097.1
132
24384
XM_017010609.1
XP_016866098.1
131
24191
XM_017010610.1
XP_016866099.1
130
24170
XM_017010611.1
XP_016866100.1
130
23973
XM_017010612.1
XP_016866101.1
129
23897
XM_017010613.1
XP_016866102.1
131
24266
XM_017010614.1
XP_016866103.1
130
24113
XM_017010615.1
XP_016866104.1
130
24113
XM_017010616.1
XP_016866105.1
131
24269
XM_017010617.1
XP_016866106.1
131
24266
XM_017010618.1
XP_016866107.1
131
24254
XM_017010619.1
XP_016866108.1
117
22348
GenBank mRNA
AB018339.2
-
19
2989
AF444779.1
-
6
862
AF495910.1
-
131
24352
AF535142.1
-
130
23974
AK122691.1
-
11
1632
AK127076.1
-
15
2329
AK304825.1
-
4
554
AK316101.1
-
4
569
AY061755.1
-
46
7442
AY061756.1
-
3
432
AY184203.1
-
2
297
AY184206.1
-
44
7337
BC090927.1
-
4
614
BX537837.1
-
13
1930
BX647837.1
-
16
2524
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nucleus organization
Golgi organization
establishment of nucleus localization
muscle cell differentiation
cytoskeletal anchoring at nuclear membrane
nuclear matrix anchoring at nuclear membrane
actin binding
protein binding
lamin binding
protein homodimerization activity
poly(A) RNA binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-9ljwx:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0