Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HBA1 SIRNA Assays ›
Gene Symbol:
Hba1
Hba2
Gene Name:
hemoglobin, alpha 1
hemoglobin, alpha 2
Gene Aliases:
HBAM, Hba-a1, Hba-a2
-
Chromosome Location:
Chr.10: 15602794 - 15603649 on Build Rnor_6.0
Chromosome Location:
Chr.10: 15589364 - 15590207 on Build Rnor_6.0
UniGene ID:
Rn.107334
Rn.203003
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID
s236166
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hba1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013096.1
NP_037228.1
1
101
GenBank mRNA
BC059150.1
-
1
100
6918464
BC091567.1
-
88
7458326
BC160818.1
-
1
84
7375173
FQ209733.1
-
1
101
FQ209744.1
-
1
101
FQ209909.1
-
1
101
FQ209912.1
-
1
101
FQ210001.1
-
1
101
FQ210150.1
-
1
99
FQ210186.1
-
1
99
FQ210277.1
-
1
99
FQ210349.1
-
1
99
FQ210635.1
-
1
101
FQ210643.1
-
1
99
FQ210690.1
-
1
100
FQ210723.1
-
1
99
FQ210862.1
-
1
99
FQ210866.1
-
1
99
FQ211077.1
-
1
101
FQ211217.1
-
1
99
FQ211220.1
-
1
101
FQ211232.1
-
1
100
FQ211239.1
-
1
99
FQ211351.1
-
1
99
FQ211374.1
-
1
99
FQ211431.1
-
1
99
FQ211513.1
-
1
99
FQ211520.1
-
1
99
FQ211755.1
-
1
99
FQ211911.1
-
1
99
FQ211972.1
-
1
99
FQ212025.1
-
1
101
FQ212144.1
-
1
101
FQ212153.1
-
1
101
FQ212186.1
-
1
99
FQ212232.1
-
1
101
FQ212457.1
-
1
99
FQ212854.1
-
1
99
FQ213301.1
-
1
99
FQ214066.1
-
1
99
FQ214223.1
-
1
99
FQ216243.1
-
1
99
FQ217723.1
-
1
99
FQ217724.1
-
1
99
FQ217869.1
-
1
99
FQ217988.1
-
1
101
FQ218117.1
-
1
99
FQ218146.1
-
99
FQ218612.1
-
1
98
FQ218968.1
-
1
101
FQ219715.1
-
1
99
FQ220786.1
-
1
99
FQ221656.1
-
1
100
FQ221912.1
-
1
101
FQ221952.1
-
1
99
FQ222431.1
-
1
99
FQ222454.1
-
1
99
FQ222672.1
-
1
99
FQ222750.1
-
1
101
FQ223017.1
-
1
100
FQ224581.1
-
1
99
FQ225427.1
-
1
100
FQ225429.1
-
1
101
FQ225440.1
-
1
100
FQ225452.1
-
1
99
FQ225777.1
-
1
99
FQ225808.1
-
1
102
FQ226118.1
-
1
101
FQ226173.1
-
1
101
FQ226175.1
-
1
99
FQ226203.1
-
1
99
FQ226615.1
-
1
99
FQ227082.1
-
1
99
FQ228301.1
-
1
99
FQ228436.1
-
99
FQ230483.1
-
1
101
FQ230901.1
-
1
99
FQ231201.1
-
1
91
FQ231290.1
-
1
99
FQ232272.1
-
1
99
FQ232481.1
-
1
99
FQ234683.1
-
1
99
M17083.1
-
1
101
Gene Symbol: Hba2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007722.1
NP_001007723.1
101
GenBank mRNA
FQ209950.1
-
1
99
FQ210108.1
-
1
99
FQ210256.1
-
1
99
FQ210264.1
-
1
99
FQ210372.1
-
1
104
FQ210420.1
-
1
99
FQ210570.1
-
1
99
FQ210694.1
-
1
99
FQ211180.1
-
1
99
FQ211270.1
-
1
99
FQ211366.1
-
1
99
FQ211672.1
-
1
99
FQ211887.1
-
1
99
FQ213126.1
-
1
99
FQ214258.1
-
1
99
FQ215087.1
-
1
99
FQ217874.1
-
1
99
FQ218140.1
-
1
101
FQ218297.1
-
1
105
FQ218784.1
-
1
98
FQ221489.1
-
1
99
FQ222342.1
-
1
99
FQ222639.1
-
1
98
FQ222693.1
-
1
97
FQ223754.1
-
1
101
FQ223838.1
-
1
125
FQ224218.1
-
1
108
FQ224339.1
-
1
101
FQ224679.1
-
1
99
FQ225311.1
-
1
99
FQ225451.1
-
1
99
FQ225711.1
-
1
99
FQ225789.1
-
1
101
FQ226185.1
-
1
99
FQ226226.1
-
254
FQ226238.1
-
1
89
FQ226556.1
-
1
99
FQ226574.1
-
1
99
FQ226576.1
-
1
99
FQ226594.1
-
1
99
FQ226619.1
-
1
99
FQ226927.1
-
1
99
FQ228017.1
-
1
101
FQ228292.1
-
1
99
FQ228298.1
-
1
99
FQ228306.1
-
1
99
FQ228588.1
-
1
99
FQ229556.1
-
1
99
FQ230499.1
-
1
99
FQ230501.1
-
1
99
FQ230536.1
-
1
99
FQ230819.1
-
1
101
FQ230868.1
-
1
99
FQ230877.1
-
1
101
FQ231222.1
-
1
99
FQ231239.1
-
1
94
FQ231246.1
-
1
99
FQ231250.1
-
1
99
FQ231268.1
-
1
98
FQ231572.1
-
1
99
FQ231902.1
-
1
99
FQ232213.1
-
1
99
FQ234805.1
-
1
99
FQ235117.1
-
1
100
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
oxygen transport
negative regulation of blood pressure
protein heterooligomerization
regulation of sensory perception of pain
in utero embryonic development
positive regulation of cell death
response to hydrogen peroxide
hydrogen peroxide catabolic process
erythrocyte development
oxidation-reduction process
beta-amyloid binding
oxygen transporter activity
iron ion binding
protein binding
oxygen binding
heme binding
organic acid binding
peroxidase activity
haptoglobin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0