Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist2h3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107698.1 | NP_001101168.1 | 1 | 310 | ||
XM_008761309.1 | XP_008759531.1 | 1 | 367 |
Gene Symbol: LOC690126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761346.1 | XP_008759568.1 | 1 | 362 |