Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138533.3 | NP_612542.1 | 6 | 1125 | ||
GenBank mRNA | BC078734.1 | - | 6 | 1125 | 7121065 |