Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VEGFA SIRNA Assays ›
Gene Symbol:
Vegfa
Gene Name:
vascular endothelial growth factor A
Gene Aliases:
VEGF-A, VEGF164, VPF, Vegf
Chromosome Location:
Chr.9: 17340341 - 17355681 on Build Rnor_6.0
UniGene ID:
Rn.1923
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s236285
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Vegfa
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001110333.2
NP_001103803.1
4, 5
1414
NM_001110334.2
NP_001103804.1
4, 5
1414
NM_001110335.1
NP_001103805.1
1, 2
469
NM_001110336.1
NP_001103806.1
1, 2
469
NM_001287107.1
NP_001274036.1
4, 5
1414
NM_001287108.1
NP_001274037.1
4, 5
1414
NM_001287110.1
NP_001274039.1
4, 5
1414
NM_001287111.1
NP_001274040.1
4, 5
1414
NM_001287112.1
NP_001274041.1
4, 5
1414
NM_001287113.1
NP_001274042.1
3, 4
1365
NM_001287114.1
NP_001274043.1
3, 4
1365
NM_031836.3
NP_114024.2
4, 5
1414
GenBank mRNA
AF062644.1
-
1, 2
65
AF215725.1
-
4, 5
400
AF215726.1
-
4, 5
400
AF222779.1
-
4, 5
400
AF260425.1
-
1, 2
68
AF261751.1
-
1, 2
69
AY033503.1
-
3, 4
362
AY033504.1
-
3, 4
362
AY033506.1
-
4, 5
411
AY033507.1
-
4, 5
411
AY033508.1
-
4, 5
411
AY702972.1
-
4, 5
400
BC168708.1
-
4, 5
1290
7458665
CK357826.1
-
1, 2
469
CK364938.1
-
1, 2
259
EU040284.1
-
1, 2
59
EU040285.1
-
1, 2
59
GQ423618.1
-
3, 4
322
L20913.1
-
2, 3
284
M32167.1
-
4, 5
404
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
ovarian follicle development
blood vessel development
patterning of blood vessels
response to hypoxia
in utero embryonic development
kidney development
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
vasculature development
lymphangiogenesis
blood vessel remodeling
sprouting angiogenesis
cell migration involved in sprouting angiogenesis
positive regulation of neuroblast proliferation
positive regulation of mesenchymal cell proliferation
positive regulation of receptor internalization
basophil chemotaxis
heart morphogenesis
outflow tract morphogenesis
coronary vein morphogenesis
regulation of transcription from RNA polymerase II promoter
nervous system development
mesoderm development
female pregnancy
aging
lactation
cell proliferation
positive regulation of cell proliferation
regulation of cell shape
response to cold
positive regulation of signal transduction
positive regulation of endothelial cell migration
positive regulation of gene expression
cell migration
negative regulation of cell-cell adhesion
cell differentiation
hyaluronan metabolic process
monocyte differentiation
macrophage differentiation
lung development
positive regulation of cell migration
regulation of cGMP metabolic process
epithelial cell differentiation
positive regulation of vascular endothelial growth factor receptor signaling pathway
post-embryonic camera-type eye development
positive regulation of protein complex assembly
positive regulation of protein autophosphorylation
activation of protein kinase activity
response to estradiol
response to progesterone
positive regulation of CREB transcription factor activity
positive regulation of peptidyl-serine phosphorylation
response to vitamin A
tube formation
cellular response to drug
endothelial cell chemotaxis
cellular response to vascular endothelial growth factor stimulus
lymph vessel morphogenesis
positive regulation of endothelial cell chemotaxis by VEGF-activated vascular endothelial growth factor receptor signaling pathway
vascular endothelial growth factor signaling pathway
positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway
VEGF-activated neuropilin signaling pathway
growth
wound healing
T-helper 1 type immune response
eye photoreceptor cell development
negative regulation of apoptotic process
negative regulation of programmed cell death
positive regulation of vascular permeability
surfactant homeostasis
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of MAP kinase activity
negative regulation of neuron apoptotic process
positive regulation of blood vessel endothelial cell migration
endothelial cell migration
cellular response to fibroblast growth factor stimulus
regulation of endothelial cell differentiation
positive regulation of angiogenesis
negative regulation of bone resorption
positive regulation of cell adhesion
positive regulation of transcription from RNA polymerase II promoter
vascular endothelial growth factor receptor signaling pathway
mRNA stabilization
lung alveolus development
cell maturation
blood vessel morphogenesis
camera-type eye morphogenesis
positive regulation of smooth muscle cell proliferation
neuron development
cardiac muscle fiber development
branching morphogenesis of an epithelial tube
positive regulation of axon extension involved in axon guidance
artery morphogenesis
positive regulation of epithelial cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
positive regulation of positive chemotaxis
induction of positive chemotaxis
positive regulation of cellular component movement
response to folic acid
positive regulation of cell division
positive regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
primitive erythrocyte differentiation
cell chemotaxis
lung vasculature development
mammary gland alveolus development
positive regulation of mast cell chemotaxis
cardiac vascular smooth muscle cell development
angiogenesis involved in coronary vascular morphogenesis
coronary artery morphogenesis
positive regulation of transcription from RNA polymerase II promoter in response to hypoxia
positive regulation of ERK1 and ERK2 cascade
response to oxygen levels
cellular response to hypoxia
dopaminergic neuron differentiation
response to dexamethasone
cellular response to dexamethasone stimulus
commissural neuron axon guidance
positive regulation of protein kinase C signaling
positive regulation of cell migration involved in sprouting angiogenesis
positive regulation of branching involved in ureteric bud morphogenesis
regulation of retinal ganglion cell axon guidance
positive regulation of peptidyl-tyrosine autophosphorylation
positive regulation of p38MAPK cascade
negative regulation of neuron death
positive regulation of lymphangiogenesis
regulation of hematopoietic progenitor cell differentiation
positive regulation of histone deacetylase activity
positive regulation of retinal ganglion cell axon guidance
positive regulation of protein localization to early endosome
positive regulation of protein kinase D signaling
negative regulation of extrinsic apoptotic signaling pathway
fibronectin binding
receptor binding
cytokine activity
platelet-derived growth factor receptor binding
vascular endothelial growth factor receptor binding
growth factor activity
heparin binding
neuropilin binding
chemoattractant activity
identical protein binding
protein homodimerization activity
vascular endothelial growth factor receptor 1 binding
vascular endothelial growth factor receptor 2 binding
protein heterodimerization activity
receptor agonist activity
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-hjwgp:80/10.239.166.163:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0