Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vegfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110333.2 | NP_001103803.1 | 4 | 1388 | ||
NM_001110334.2 | NP_001103804.1 | 4 | 1388 | |||
NM_001110335.1 | NP_001103805.1 | 1 | 443 | |||
NM_001110336.1 | NP_001103806.1 | 1 | 443 | |||
NM_001287107.1 | NP_001274036.1 | 4 | 1388 | |||
NM_001287108.1 | NP_001274037.1 | 4 | 1388 | |||
NM_001287110.1 | NP_001274039.1 | 4 | 1388 | |||
NM_001287111.1 | NP_001274040.1 | 4 | 1388 | |||
NM_001287112.1 | NP_001274041.1 | 4 | 1388 | |||
NM_001287113.1 | NP_001274042.1 | 3 | 1339 | |||
NM_001287114.1 | NP_001274043.1 | 3 | 1339 | |||
NM_031836.3 | NP_114024.2 | 4 | 1388 | |||
GenBank mRNA | AF062644.1 | - | 1 | 39 | ||
AF215725.1 | - | 4 | 374 | |||
AF215726.1 | - | 4 | 374 | |||
AF222779.1 | - | 4 | 374 | |||
AF260425.1 | - | 1 | 42 | |||
AF261751.1 | - | 1 | 43 | |||
AY033503.1 | - | 3 | 336 | |||
AY033504.1 | - | 3 | 336 | |||
AY033506.1 | - | 4 | 385 | |||
AY033507.1 | - | 4 | 385 | |||
AY033508.1 | - | 4 | 385 | |||
AY702972.1 | - | 4 | 374 | |||
BC168708.1 | - | 4 | 1264 | 7458665 | ||
CK357826.1 | - | 1 | 443 | |||
CK364938.1 | - | 1 | 233 | |||
EU040284.1 | - | 1 | 33 | |||
EU040285.1 | - | 1 | 33 | |||
GQ423618.1 | - | 3 | 296 | |||
L20913.1 | - | 2 | 258 | |||
M32167.1 | - | 4 | 378 |