Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130965.2 | NP_001124437.1 | 9 | 986 | ||
NM_001171944.1 | NP_001165415.1 | 7 | 736 | |||
NM_025154.5 | NP_079430.3 | 8 | 849 | |||
GenBank mRNA | AB018353.1 | - | 10 | 1084 | ||
AB648918.1 | - | 11 | 1275 | |||
AK022816.1 | - | 8 | 841 | |||
AK025737.1 | - | 6 | 511 | |||
AK302896.1 | - | 7 | 708 | |||
AK304150.1 | - | 5 | 410 | |||
BC142707.1 | - | 9 | 996 | 40148817 | ||
BC152418.1 | - | 9 | 1084 | 8862528 | ||
BX640866.1 | - | 1 | 619 | |||
BX648893.1 | - | 1 | 619 | |||
BX649069.1 | - | 7 | 741 |