Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130965.2 | NP_001124437.1 | 12, 13 | 1416 | ||
NM_001171944.1 | NP_001165415.1 | 10, 11 | 1166 | |||
NM_025154.5 | NP_079430.3 | 11, 12 | 1279 | |||
GenBank mRNA | AB018353.1 | - | 13, 14 | 1514 | ||
AB648918.1 | - | 14, 15 | 1705 | |||
AF202724.1 | - | 3, 4 | 315 | |||
AK022816.1 | - | 11, 12 | 1271 | |||
AK025737.1 | - | 9, 10 | 941 | |||
AK302896.1 | - | 10, 11 | 1138 | |||
AK304150.1 | - | 8, 9 | 807 | |||
BC142707.1 | - | 12, 13 | 1426 | 40148817 | ||
BC152418.1 | - | 12, 13 | 1514 | 8862528 | ||
BX640866.1 | - | 3, 4 | 1505 | |||
BX648893.1 | - | 3, 4 | 1505 | |||
BX649069.1 | - | 10, 11 | 1171 |