Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1ql3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109403.2 | NP_001102873.1 | 2 | 685 | ||
XM_008771889.1 | XP_008770111.1 | 4 | 5144 | |||
XM_008771890.1 | XP_008770112.1 | 5 | 8202 | |||
XM_008771891.1 | XP_008770113.1 | 4 | 4467 |