Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013103.2 | NP_001013121.2 | 5 | 650 | ||
GenBank mRNA | FQ228127.1 | - | 2 | 179 |