Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109198.1 | NP_001102668.1 | 5 | 404 | ||
GenBank mRNA | BC166903.1 | - | 5 | 405 | 6886805 | |
FQ209969.1 | - | 3 | 188 | |||
FQ211114.1 | - | 3 | 188 | |||
FQ218073.1 | - | 3 | 188 | |||
FQ220669.1 | - | 3 | 188 | |||
FQ220753.1 | - | 3 | 188 | |||
FQ221328.1 | - | 3 | 188 | |||
FQ221528.1 | - | 3 | 188 | |||
FQ221617.1 | - | 3 | 188 | |||
FQ221651.1 | - | 3 | 188 | |||
FQ222328.1 | - | 3 | 188 | |||
FQ222394.1 | - | 3 | 188 | |||
FQ222706.1 | - | 3 | 188 | |||
FQ223807.1 | - | 3 | 188 | |||
FQ224022.1 | - | 3 | 188 | |||
FQ224617.1 | - | 3 | 188 | |||
FQ227863.1 | - | 3 | 188 | |||
FQ228326.1 | - | 3 | 188 | |||
FQ228502.1 | - | 3 | 188 | |||
FQ228613.1 | - | 3 | 188 | |||
FQ231502.1 | - | 3 | 188 |