Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214457.2 | NP_999622.2 | 13 | 1866 | ||
GenBank mRNA | AY494977.1 | - | 1735 | |||
BC098634.1 | - | 13 | 1866 | 7317963 |