Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpv17l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098240.1 | NP_001091710.1 | 5 | 649 | ||
XM_008764515.1 | XP_008762737.1 | 5 | 5497 | |||
XM_008764516.1 | XP_008762738.1 | 5 | 5496 | |||
XM_008764518.1 | XP_008762740.1 | 5 | 638 | |||
XM_008764519.1 | XP_008762741.1 | 6 | 475 | |||
GenBank mRNA | BC091193.1 | - | 5 | 649 | 7373705 |