Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cobll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234301.2 | XP_006234363.1 | 13 | 5439 | ||
XM_006234304.2 | XP_006234366.1 | 12 | 5333 | |||
XM_008761934.1 | XP_008760156.1 | 13 | 4245 | |||
XM_008761935.1 | XP_008760157.1 | 13 | 4230 | |||
XM_008761936.1 | XP_008760158.1 | 12 | 4206 | |||
XM_008761937.1 | XP_008760159.1 | 13 | 4245 | |||
XM_008761938.1 | XP_008760160.1 | 14 | 5480 | |||
XM_008761939.1 | XP_008760161.1 | 12 | 4140 | |||
XM_008761940.1 | XP_008760162.1 | 12 | 4125 | |||
XM_008761941.1 | XP_008760163.1 | 13 | 5441 | |||
XM_008761942.1 | XP_008760164.1 | 13 | 5833 | |||
XM_008761943.1 | XP_008760165.1 | 12 | 5335 | |||
GenBank mRNA | BC168937.1 | - | 1 | 712 |