Display Info New header:
tfpt-header-footer.tfcom-eks-na-comerce-gateway-prd.commerceprod.thermofisher.net cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@201e6afb
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous enableSimpleHeaderValue:
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Special Offers
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search All
Popular Product Areas
TaqMan Assays
Gene Expression
Microbe & ABR Detection
Copy Number Variation
Mutation Detection
miRNA
SNP Genotyping
Primary Antibodies
Secondary Antibodies
Isotype Controls
Proteins & Peptides
ELISA Kits
All Documents & Support
Certificates
SDS
Manuals & Protocols
Product FAQs
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HMGB1 SIRNA Assays ›
Gene Symbol:
Hmgb1
Gene Name:
high mobility group box 1
Gene Aliases:
Ac2-008, Hmg1
Chromosome Location:
Chr.12: 7090927 - 7092190 on Build Rnor_6.0
UniGene ID:
Rn.144565
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s236588
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
40 nmol
InVivo | 5 nmol
20 nmol
40 nmol
250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
10 umol
250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
10 umol
Availability
Made To Order
Catalog #
4390771
Price (USD)
400.00
Your Price
Online offer (USD):
388.65
388.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
Displaying...
Build 6.0 Rat Chr.12: s236588
×
Created with Raphaël 2.1.2
Hmgb1
7,087,00
0
7,087,10
0
7,087,20
0
7,087,30
0
7,087,40
0
7,087,50
0
7,087,60
0
7,087,70
0
7,087,80
0
7,087,90
0
7,088,00
0
7,088,10
0
7,088,20
0
7,088,30
0
7,088,40
0
7,088,50
0
7,088,60
0
7,088,70
0
7,088,80
0
7,088,90
0
7,089,00
0
7,089,10
0
7,089,20
0
7,089,30
0
7,089,40
0
7,089,50
0
7,089,60
0
7,089,70
0
7,089,80
0
7,089,90
0
7,090,00
0
7,090,10
0
7,090,20
0
7,090,30
0
7,090,40
0
7,090,50
0
7,090,60
0
7,090,70
0
7,090,80
0
7,090,90
0
7,091,00
0
7,091,10
0
7,091,20
0
7,091,30
0
7,091,40
0
7,091,50
0
7,091,60
0
7,091,70
0
7,091,80
0
7,091,90
0
7,092,00
0
7,092,10
0
7,092,20
0
7,092,30
0
7,092,40
0
7,092,50
0
7,092,60
0
7,092,70
0
7,092,80
0
7,092,90
0
7,093,00
0
7,093,10
0
7,093,20
0
7,093,30
0
7,093,40
0
7,093,50
0
7,093,60
0
7,093,70
0
7,093,80
0
7,093,90
0
7,094,00
0
7,094,10
0
7,094,20
0
7,094,30
0
7,094,40
0
7,094,50
0
7,094,60
0
7,094,70
0
7,094,80
0
7,094,90
0
7,095,00
0
7,095,10
0
7,095,20
0
7,095,30
0
7,095,40
0
7,095,50
0
7,095,60
0
7,095,70
0
7,095,80
0
7,095,90
0
7,096,00
0
7,096,10
0
Show all RefSeq and GenBank
Back To Top
siRNA Details
Gene Symbol: Hmgb1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012963.2
NP_037095.1
1890
GenBank mRNA
BC081839.1
-
1890
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
cell morphogenesis
eye development
positive regulation of protein phosphorylation
positive regulation of mesenchymal cell proliferation
inflammatory response to antigenic stimulus
base-excision repair, DNA ligation
chromatin remodeling
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
chemotaxis
nervous system development
circadian rhythm
negative regulation of DNA replication
positive regulation of cell proliferation
response to heat
response to glucose
positive regulation of cell death
positive regulation of neuron projection development
positive regulation of smooth muscle cell migration
negative regulation of RNA polymerase II transcriptional preinitiation complex assembly
lung development
positive regulation of cell migration
neuron projection development
actin cytoskeleton reorganization
DNA geometric change
response to lipopolysaccharide
response to insulin
positive regulation of myeloid cell apoptotic process
V(D)J recombination
response to interferon-gamma
response to drug
positive regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of DNA binding
positive regulation of myeloid cell differentiation
positive regulation of myoblast differentiation
positive regulation of glycogen catabolic process
positive regulation of protein kinase activity
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
regulation of inflammatory response
male-specific defense response to bacterium
induction of positive chemotaxis
response to glucocorticoid
myoblast proliferation
cellular response to interleukin-1
four-way junction DNA binding
open form four-way junction DNA binding
crossed form four-way junction DNA binding
DNA binding
bent DNA binding
chromatin binding
double-stranded DNA binding
single-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
cytokine activity
protein binding
5S rRNA binding
transcription factor binding
heparin binding
DNA binding, bending
calcium-dependent protein kinase regulator activity
protein kinase activator activity
peptide binding
poly(A) RNA binding
protein dimerization activity
RAGE receptor binding
glycolipid binding
repressing transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-59d6677d6-zmlwt:80/100.66.78.27:80.
git-commit: 1e3318a2800c8d0a8829a508293490082e5b9833
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.30.0-Offline