Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134993.1 | NP_001128465.1 | 7 | 1480 | ||
GenBank mRNA | BC169052.1 | - | 7 | 1444 | 7386405 |