Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rpl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077592.1 | NP_001071060.1 | 5 | 289 | ||
XM_008768393.1 | XP_008766615.1 | 5 | 288 | |||
GenBank mRNA | BC101933.1 | - | 5 | 289 | 7371608 | |
FQ210073.1 | - | 5 | 296 | |||
FQ217659.1 | - | 5 | 291 | |||
FQ222790.1 | - | 5 | 296 | |||
FQ223953.1 | - | 5 | 296 | |||
FQ224522.1 | - | 5 | 296 | |||
FQ228832.1 | - | 5 | 307 | |||
X57007.1 | - | 5 | 294 |
Gene Symbol: LOC685963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001065954.5 | XP_001065954.1 | 1 | 311 |
Gene Symbol: LOC690468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001074532.4 | XP_001074532.1 | 1 | 293 |