Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025746.3 | NP_001020917.3 | 4, 5 | 1376 | ||
GenBank mRNA | DQ058650.1 | - | 4, 5 | 1376 |
Gene Symbol: Rhox7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025654.1 | NP_001020825.1 | 4, 5 | 651 | ||
GenBank mRNA | DQ058654.1 | - | 4, 5 | 651 |