Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMK2G SIRNA Assays ›
Gene Symbol:
CAMK2G
Gene Name:
calcium/calmodulin dependent protein kinase II gamma
Gene Aliases:
CAMK, CAMK-II, CAMKG
Chromosome Location:
Chr.10: 73812501 - 73874591 on Build GRCh38
UniGene ID:
Hs.523045
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2368
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMK2G
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204492.1
NP_001191421.1
3
302
NM_001222.3
NP_001213.2
3
302
NM_001320898.1
NP_001307827.1
3
302
NM_172169.2
NP_751909.1
3
302
NM_172170.4
NP_751910.1
3
302
NM_172171.2
NP_751911.1
3
302
NM_172173.2
NP_751913.1
3
302
XM_005270195.1
XP_005270252.1
3
293
XM_005270197.1
XP_005270254.1
3
293
XM_005270198.1
XP_005270255.1
3
293
XM_005270199.1
XP_005270256.1
3
293
XM_005270200.1
XP_005270257.1
3
293
XM_005270201.1
XP_005270258.1
3
293
XM_005270203.1
XP_005270260.1
3
293
XM_005270205.1
XP_005270262.1
3
293
XM_006717993.1
XP_006718056.1
3
293
XM_006717996.1
XP_006718059.1
3
293
XM_006717997.1
XP_006718060.1
3
293
XM_006717998.1
XP_006718061.1
3
293
XM_011540225.2
XP_011538527.1
3
241
XM_017016716.1
XP_016872205.1
3
293
XM_017016717.1
XP_016872206.1
3
241
XM_017016718.1
XP_016872207.1
3
293
XM_017016719.1
XP_016872208.1
3
293
XM_017016720.1
XP_016872209.1
3
293
XM_017016721.1
XP_016872210.1
3
368
XM_017016722.1
XP_016872211.1
3
293
XM_017016723.1
XP_016872212.1
3
293
XM_017016724.1
XP_016872213.1
3
293
XM_017016725.1
XP_016872214.1
3
293
XM_017016726.1
XP_016872215.1
3
293
XM_017016727.1
XP_016872216.1
3
293
XM_017016728.1
XP_016872217.1
3
241
XM_017016729.1
XP_016872218.1
2
83
XM_017016730.1
XP_016872219.1
3
293
XM_017016731.1
XP_016872220.1
3
254
XM_017016732.1
XP_016872221.1
3
187
XM_017016733.1
XP_016872222.1
3
241
XM_017016734.1
XP_016872223.1
3
280
XM_017016735.1
XP_016872224.1
3
293
XM_017016736.1
XP_016872225.1
3
368
XM_017016737.1
XP_016872226.1
3
250
XM_017016738.1
XP_016872227.1
3
254
XM_017016739.1
XP_016872228.1
3
425
GenBank mRNA
AK095576.1
-
4
399
AK131199.1
-
3
225
AK225771.1
-
3
302
AK291704.1
-
3
294
AK301180.1
-
3
294
BC034044.1
-
3
293
4813948
BU165166.1
-
3
299
DR002605.1
-
3
309
L07044.1
-
3
193
U66064.1
-
3
225
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
MAPK cascade
protein dephosphorylation
calcium ion transport
nervous system development
regulation of skeletal muscle adaptation
insulin secretion
cell differentiation
positive regulation of GTPase activity
protein autophosphorylation
protein oligomerization
regulation of calcium ion transport
interferon-gamma-mediated signaling pathway
regulation of cellular response to heat
regulation of relaxation of cardiac muscle
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
calcium-dependent protein serine/threonine phosphatase activity
Ras guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0