Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134706.1 | NP_001128178.1 | 6 | 720 | ||
GenBank mRNA | BC168165.1 | - | 6 | 622 | 8370295 |
Gene Symbol: Hmgn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276434.1 | NP_001263363.1 | 6 | 720 | ||
GenBank mRNA | DY318327.1 | - | 6 | 619 |