Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hist1h2ak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109423.1 | NP_001102893.1 | 1 | 274 |
Gene Symbol: H2afx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109291.1 | NP_001102761.1 | 1 | 322 | ||
GenBank mRNA | CB713471.1 | - | 1 | 322 |
Gene Symbol: Hist1h2aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021839.1 | NP_068611.1 | 1 | 274 |
Gene Symbol: Hist1h2ac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751712.3 | XP_003751760.1 | 1 | 320 |
Gene Symbol: Hist1h2ai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_577577.6 | XP_577577.1 | 1 | 290 |
Gene Symbol: Hist3h2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021840.2 | NP_068612.2 | 1 | 279 | ||
GenBank mRNA | BC099140.1 | - | 1 | 279 | 7457173 |
Gene Symbol: LOC100360754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728479.4 | XP_002728525.1 | 1 | 319 |
Gene Symbol: LOC100910554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751711.3 | XP_003751759.1 | 1 | 362 |
Gene Symbol: LOC103690190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771771.1 | XP_008769993.1 | 1 | 319 |
Gene Symbol: LOC680322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771767.1 | XP_008769989.1 | 1 | 394 |