Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h2ak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109423.1 | NP_001102893.1 | 1 | 217 |
Gene Symbol: H2afj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109610.2 | NP_001103080.1 | 1 | 251 | ||
GenBank mRNA | BC157816.1 | - | 1 | 251 | 9034669 |
Gene Symbol: Hist1h2ac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751712.3 | XP_003751760.1 | 1 | 263 |
Gene Symbol: Hist1h2ah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253977.2 | XP_006254039.1 | 1 | 278 |
Gene Symbol: LOC103690002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763447.1 | XP_008761669.1 | 1 | 290 |
Gene Symbol: LOC680322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771767.1 | XP_008769989.1 | 1 | 337 |