Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC690460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109593.1 | NP_001103063.1 | 1 | 415 |
Gene Symbol: LOC687631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772838.1 | XP_008771060.1 | 1 | 566 |
Gene Symbol: LOC689927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109555.1 | NP_001103025.1 | 496 |
Gene Symbol: LOC690431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772842.1 | XP_008771064.1 | 2 | 448 | ||
XM_008772843.1 | XP_008771065.1 | 2 | 436 | |||
XM_008772844.1 | XP_008771066.1 | 2 | 415 | |||
XM_008772845.1 | XP_008771067.1 | 2 | 415 |