Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134705.1 | NP_001128177.1 | 2 | 300 | ||
GenBank mRNA | BC166894.1 | - | 2 | 264 | 6920183 | |
FQ217721.1 | - | 2 | 249 | |||
FQ217795.1 | - | 2 | 249 | |||
FQ221226.1 | - | 2 | 310 | |||
FQ223619.1 | - | 2 | 251 | |||
FQ224715.1 | - | 2 | 279 |