Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278731.1 | NP_001265660.1 | 11 | 1548 | ||
NM_001278732.1 | NP_001265661.1 | 10 | 1501 | |||
NM_012208.3 | NP_036340.1 | 12 | 1623 | |||
XM_011537619.1 | XP_011535921.1 | 13 | 1636 | |||
XM_011537620.1 | XP_011535922.1 | 12 | 1537 | |||
XM_017009287.1 | XP_016864776.1 | 11 | 1524 | |||
XM_017009288.1 | XP_016864777.1 | 13 | 1687 | |||
XM_017009289.1 | XP_016864778.1 | 13 | 1680 | |||
XM_017009290.1 | XP_016864779.1 | 12 | 1747 | |||
XM_017009291.1 | XP_016864780.1 | 12 | 1440 | |||
XM_017009292.1 | XP_016864781.1 | 12 | 1384 | |||
GenBank mRNA | AK293269.1 | - | 10 | 1281 | ||
AK293390.1 | - | 11 | 1365 | |||
AK298661.1 | - | 7 | 919 | |||
AK299971.1 | - | 9 | 1077 | |||
AK312976.1 | - | 12 | 1441 | |||
BC007680.2 | - | 12 | 1442 | 3628006 | ||
BC014982.1 | - | 12 | 1587 | 4859849 | ||
U18937.1 | - | 12 | 1549 |