Gene Symbol: PHF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290259.1 | NP_001277188.1 | 17 | 4654 | ||
NM_015153.3 | NP_055968.1 | 15 | 4113 | |||
XM_005248701.3 | XP_005248758.1 | 16 | 4448 | |||
XM_005248702.3 | XP_005248759.1 | 15 | 4192 | |||
XM_005248703.3 | XP_005248760.1 | 14 | 2402 | |||
XM_006715426.3 | XP_006715489.1 | 16 | 4673 | |||
XM_011535648.2 | XP_011533950.1 | 16 | 4330 | |||
XM_017010626.1 | XP_016866115.1 | 15 | 2665 | |||
GenBank mRNA | AF091622.1 | - | 15 | 4114 | ||
AK025283.1 | - | 1 | 46 | |||
AK298580.1 | - | 14 | 2386 | |||
BC113650.1 | - | 15 | 4114 | 8322702 | ||
BC113652.1 | - | 15 | 4114 | 8322704 | ||
BX648268.1 | - | 17 | 4340 | |||
D87685.2 | - | 15 | 4102 |