Gene Symbol: PHF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290259.1 | NP_001277188.1 | 5 | 1852 | ||
NM_001290260.1 | NP_001277189.1 | 4 | 1652 | |||
NM_015153.3 | NP_055968.1 | 3 | 1311 | |||
XM_005248701.3 | XP_005248758.1 | 4 | 1646 | |||
XM_005248702.3 | XP_005248759.1 | 3 | 1390 | |||
XM_006715426.3 | XP_006715489.1 | 4 | 1871 | |||
XM_011535648.2 | XP_011533950.1 | 4 | 1528 | |||
GenBank mRNA | AF091622.1 | - | 3 | 1312 | ||
AK055678.1 | - | 4 | 1567 | |||
AK290491.1 | - | 4 | 1338 | |||
AK307869.1 | - | 2 | 1211 | |||
AK315868.1 | - | 3 | 1370 | |||
BC113650.1 | - | 3 | 1312 | 8322702 | ||
BC113652.1 | - | 3 | 1312 | 8322704 | ||
BX648268.1 | - | 5 | 1538 | |||
D87685.2 | - | 3 | 1300 |