Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CAPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014296.2 | NP_055111.1 | 17, 18 | 2273 | ||
XM_011533555.2 | XP_011531857.1 | 18, 19 | 2172 | |||
XM_011533556.2 | XP_011531858.1 | 19, 20 | 2404 | |||
XM_017006043.1 | XP_016861532.1 | 18, 19 | 2356 | |||
GenBank mRNA | AB028639.1 | - | 17, 18 | 2254 | ||
AK314253.1 | - | 17, 18 | 2235 | |||
BC056202.1 | - | 17, 18 | 2251 | 5274889 |