Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014296.2 | NP_055111.1 | 19 | 2452 | ||
XM_011533555.2 | XP_011531857.1 | 20 | 2351 | |||
XM_011533556.2 | XP_011531858.1 | 21 | 2583 | |||
XM_011533557.2 | XP_011531859.1 | 18 | 2191 | |||
XM_017006043.1 | XP_016861532.1 | 20 | 2535 | |||
XM_017006044.1 | XP_016861533.1 | 17 | 2143 | |||
XM_017006045.1 | XP_016861534.1 | 18 | 2209 | |||
GenBank mRNA | AB028639.1 | - | 19 | 2433 | ||
AK093742.1 | - | 2 | 1017 | |||
AK314253.1 | - | 19 | 2414 | |||
BC056202.1 | - | 19 | 2430 | 5274889 |