Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134476.1 | NP_001127948.1 | 7 | 2537 | ||
NM_015350.2 | NP_056165.1 | 5 | 2594 | |||
XM_005270701.4 | XP_005270758.1 | 5 | 2463 | |||
XM_005270702.4 | XP_005270759.1 | 6 | 2484 | |||
XM_005270703.4 | XP_005270760.1 | 5 | 4231 | |||
XM_011541143.2 | XP_011539445.1 | 6 | 2521 | |||
XM_011541144.2 | XP_011539446.1 | 7 | 2833 | |||
XM_011541145.2 | XP_011539447.1 | 8 | 2858 | |||
XM_011541146.2 | XP_011539448.1 | 6 | 4288 | |||
XM_011541148.2 | XP_011539450.1 | 6 | 2509 | |||
XM_017000883.1 | XP_016856372.1 | 6 | 3483 | |||
XM_017000884.1 | XP_016856373.1 | 5 | 4385 | |||
XM_017000885.1 | XP_016856374.1 | 5 | 2555 | |||
XM_017000886.1 | XP_016856375.1 | 5 | 2459 | |||
GenBank mRNA | AF385436.1 | - | 5 | 2594 | ||
AY358112.1 | - | 4 | 2342 | |||
BC030607.2 | - | 7 | 2773 | 4824652 | ||
BC060782.1 | - | 7 | 2479 | 5298560 | ||
D86984.2 | - | 5 | 2900 |