Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NUP188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015354.2 | NP_056169.1 | 38 | 4390 | ||
GenBank mRNA | AK025292.1 | - | 5 | 715 | ||
BC005407.1 | - | 19 | 2331 | |||
BC040352.1 | - | 25 | 2997 | |||
BC111045.1 | - | 16 | 2039 | |||
BX537774.1 | - | 35 | 4034 | |||
D79991.1 | - | 38 | 4352 |