Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPIDR SIRNA Assays ›
Gene Symbol:
SPIDR
Gene Name:
scaffolding protein involved in DNA repair
Gene Aliases:
KIAA0146
Chromosome Location:
Chr.8: 47260878 - 47736001 on Build GRCh38
UniGene ID:
Hs.381058
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23973
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPIDR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080394.3
NP_001073863.1
11
1657
NM_001282916.1
NP_001269845.1
10
1590
NM_001282919.1
NP_001269848.1
11
1620
NR_104581.1
8
1054
XM_005251189.3
XP_005251246.1
11
1599
XM_005251191.3
XP_005251248.1
11
1599
XM_005251193.3
XP_005251250.1
11
1599
XM_005251195.4
XP_005251252.1
10
1443
XM_005251198.4
XP_005251255.1
10
1377
XM_005251199.4
XP_005251256.1
8
1191
XM_006716443.3
XP_006716506.1
11
1567
XM_006716444.3
XP_006716507.1
10
1340
XM_011517497.2
XP_011515799.1
11
1599
XM_011517500.2
XP_011515802.1
11
1497
XM_011517507.2
XP_011515809.1
3
188
XM_017013268.1
XP_016868757.1
11
1599
XM_017013269.1
XP_016868758.1
11
1599
XM_017013270.1
XP_016868759.1
11
1497
XM_017013271.1
XP_016868760.1
11
1714
XM_017013272.1
XP_016868761.1
7
1343
XM_017013273.1
XP_016868762.1
9
1157
XM_017013274.1
XP_016868763.1
10
1443
XM_017013275.1
XP_016868764.1
7
1195
XM_017013276.1
XP_016868765.1
7
1182
XM_017013277.1
XP_016868766.1
11
1584
XM_017013278.1
XP_016868767.1
10
1409
XM_017013279.1
XP_016868768.1
11
1587
XM_017013280.1
XP_016868769.1
10
1377
XM_017013281.1
XP_016868770.1
10
1342
XM_017013282.1
XP_016868771.1
10
1443
XM_017013283.1
XP_016868772.1
11
1587
XM_017013284.1
XP_016868773.1
10
1443
XM_017013285.1
XP_016868774.1
10
1377
XM_017013286.1
XP_016868775.1
8
1191
XM_017013287.1
XP_016868776.1
9
1209
XM_017013288.1
XP_016868777.1
6
961
XM_017013289.1
XP_016868778.1
7
1097
XM_017013290.1
XP_016868779.1
8
1191
XM_017013291.1
XP_016868780.1
12
1622
GenBank mRNA
AK026929.1
-
4
621
AK055680.1
-
4
219
AK122820.1
-
12
1955
AK293809.1
-
11
1580
AK294248.1
-
7
1018
AK294275.1
-
10
1512
AK301677.1
-
2
94
AK303140.1
-
1
299
AK303579.1
-
11
1567
AK309842.1
-
7
881
AK310584.1
-
2
110
BC031627.1
-
8
979
D63480.1
-
11
1585
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
cellular response to DNA damage stimulus
regulation of double-strand break repair via homologous recombination
positive regulation of protein complex assembly
regulation of establishment of protein localization to chromosome
cellular response to ionizing radiation
cellular response to hydroxyurea
cellular response to camptothecin
positive regulation of double-strand break repair
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline