Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281293.1 | NP_001268222.1 | 2 | 1972 | ||
NM_001281294.1 | NP_001268223.1 | 3 | 1864 | |||
NM_012482.4 | NP_036614.1 | 2 | 1968 | |||
XM_017000887.1 | XP_016856376.1 | 5 | 3353 | |||
XM_017000888.1 | XP_016856377.1 | 5 | 2311 | |||
GenBank mRNA | AF125158.1 | - | 1 | 1864 | ||
AJ132591.1 | - | 1 | 1385 | |||
AJ132592.1 | - | 2 | 1948 | |||
AK021707.1 | - | 1 | 271 | |||
AK022916.1 | - | 2 | 1899 | |||
AK022921.1 | - | 1 | 598 | |||
AK094701.1 | - | 3 | 1799 | |||
AK297233.1 | - | 3 | 1822 | |||
BC051905.1 | - | 1 | 1857 | 6165292 | ||
BC060820.1 | - | 2 | 1948 | 30344180 |