Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBFOX2 SIRNA Assays ›
Gene Symbol:
RBFOX2
Gene Name:
RNA binding protein, fox-1 homolog 2
Gene Aliases:
FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
Chromosome Location:
Chr.22: 35738736 - 36028892 on Build GRCh38
UniGene ID:
Hs.282998
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24047
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBFOX2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031695.2
NP_001026865.1
6, 7
971
NM_001082576.1
NP_001076045.1
6, 7
971
NM_001082577.1
NP_001076046.1
6, 7
971
NM_001082578.1
NP_001076047.1
7, 8
931
NM_001082579.1
NP_001076048.1
7, 8
928
NM_014309.2
NP_055124.1
6, 7
974
XM_005261428.2
XP_005261485.1
7, 8
1286
XM_005261429.2
XP_005261486.1
7, 8
1283
XM_005261430.2
XP_005261487.1
7, 8
1286
XM_005261431.2
XP_005261488.1
7, 8
1286
XM_005261432.2
XP_005261489.1
7, 8
1283
XM_005261433.2
XP_005261490.1
7, 8
1286
XM_005261435.1
XP_005261492.1
6, 7
975
XM_005261437.1
XP_005261494.1
6, 7
978
XM_006724185.2
XP_006724248.1
7, 8
1286
XM_006724186.2
XP_006724249.1
7, 8
1283
XM_006724187.2
XP_006724250.1
7, 8
1286
XM_006724188.2
XP_006724251.1
7, 8
1286
XM_006724189.3
XP_006724252.1
7, 8
979
XM_006724190.1
XP_006724253.1
7, 8
673
XM_006724191.1
XP_006724254.1
6, 7
978
XM_006724192.2
XP_006724255.1
6, 7
808
XM_006724193.3
XP_006724256.1
7, 8
1286
XM_006724194.2
XP_006724257.1
6, 7
750
XM_017028686.1
XP_016884175.1
7, 8
665
XM_017028687.1
XP_016884176.1
7, 8
846
XM_017028688.1
XP_016884177.1
7, 8
1283
XM_017028689.1
XP_016884178.1
7, 8
751
XM_017028690.1
XP_016884179.1
6, 7
728
XM_017028691.1
XP_016884180.1
6, 7
725
XM_017028692.1
XP_016884181.1
7, 8
1020
XM_017028693.1
XP_016884182.1
6, 7
978
XM_017028694.1
XP_016884183.1
6, 7
975
XM_017028695.1
XP_016884184.1
7, 8
735
XM_017028696.1
XP_016884185.1
6, 7
978
XM_017028697.1
XP_016884186.1
7, 8
1755
XM_017028698.1
XP_016884187.1
6, 7
975
GenBank mRNA
AB649123.1
-
6, 7
606
AF229058.1
-
6, 7
776
AK055213.1
-
7, 8
892
AK291460.1
-
6, 7
855
AK314632.1
-
6, 7
790
AL009266.1
-
6, 7
779
AM419009.1
-
6, 7
630
AY072786.1
-
7, 8
741
AY960684.1
-
6, 7
606
BC013115.1
-
5, 6
741
4346741
BC025281.1
-
6, 7
794
4555865
CR456559.1
-
6, 7
672
DQ778625.1
-
7, 8
819
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
fibroblast growth factor receptor signaling pathway
regulation of definitive erythrocyte differentiation
RNA metabolic process
radial glia guided migration of Purkinje cell
intracellular estrogen receptor signaling pathway
regulation of cell proliferation
negative regulation of transcription, DNA-templated
dendrite morphogenesis
neuromuscular process controlling balance
nucleotide binding
transcription corepressor activity
RNA binding
mRNA binding
protein binding
transcription factor binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-lzswf:80/10.239.160.35:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0