Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319116.1 | NP_001306045.1 | 12 | 1246 | ||
NM_001319117.1 | NP_001306046.1 | 11 | 1138 | |||
NM_001319118.1 | NP_001306047.1 | 12 | 1174 | |||
NM_001319119.1 | NP_001306048.1 | 12 | 1454 | |||
NM_001319120.1 | NP_001306049.1 | 11 | 1163 | |||
NM_001319121.1 | NP_001306050.1 | 13 | 1147 | |||
NM_001319122.1 | NP_001306051.1 | 12 | 1239 | |||
NM_012100.3 | NP_036232.2 | 12 | 1271 | |||
NR_134970.1 | 10 | 1087 | ||||
GenBank mRNA | AF005050.2 | - | 12 | 1246 | ||
AK001777.1 | - | 12 | 1271 | |||
AK001977.1 | - | 12 | 1243 | |||
AK023005.1 | - | 12 | 1185 | |||
AK301919.1 | - | 12 | 1231 | |||
BC000653.2 | - | 12 | 1153 | 3343505 | ||
BC004854.2 | - | 8 | 804 |