Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCNDBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012142.4 | NP_036274.3 | 11 | 1269 | ||
NR_027513.2 | 9 | 1092 | ||||
NR_027514.2 | 10 | 1172 | ||||
NR_045998.1 | 10 | 1189 | ||||
NR_045999.1 | 8 | 1032 | ||||
XM_006720448.1 | XP_006720511.1 | 9 | 1191 | |||
GenBank mRNA | AF082569.1 | - | 11 | 1128 | ||
AF087852.1 | - | 11 | 1114 | |||
AF113535.1 | - | 9 | 1078 | |||
AF132034.1 | - | 11 | 1084 | |||
AF246144.1 | - | 11 | 1086 | |||
AK075146.1 | - | 11 | 1431 | |||
AK075296.1 | - | 9 | 1083 | |||
AK225143.1 | - | 11 | 1084 | |||
AK225645.1 | - | 11 | 1098 | |||
AK290665.1 | - | 11 | 1098 | |||
AK290707.1 | - | 10 | 1051 | |||
AK295017.1 | - | 10 | 1001 | |||
BC009689.1 | - | 11 | 1112 | 3896421 | ||
CR450331.1 | - | 11 | 1038 |