Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ORC3 SIRNA Assays ›
Gene Symbol:
ORC3
Gene Name:
origin recognition complex subunit 3
Gene Aliases:
LAT, LATHEO, ORC3L
Chromosome Location:
Chr.6: 87590067 - 87677824 on Build GRCh38
UniGene ID:
Hs.410228
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24169
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ORC3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001197259.1
NP_001184188.1
6
653
NM_012381.3
NP_036513.2
7
751
NM_181837.2
NP_862820.1
7
751
XM_005248704.1
XP_005248761.1
7
697
XM_005248705.1
XP_005248762.1
8
755
XM_005248706.1
XP_005248763.1
8
754
XM_011535651.1
XP_011533953.1
7
697
XM_011535652.2
XP_011533954.1
7
697
XM_017010632.1
XP_016866121.1
7
697
XM_017010633.1
XP_016866122.1
7
697
XM_017010634.1
XP_016866123.1
7
697
XM_017010635.1
XP_016866124.1
7
697
XM_017010636.1
XP_016866125.1
8
755
XM_017010637.1
XP_016866126.1
8
754
XM_017010638.1
XP_016866127.1
8
755
XM_017010639.1
XP_016866128.1
8
754
XM_017010640.1
XP_016866129.1
8
754
GenBank mRNA
AF093535.1
-
7
675
AF125507.1
-
7
684
AF135044.1
-
7
649
AK222795.1
-
7
677
AK302750.1
-
8
745
AK303056.1
-
7
677
AK303182.1
-
6
472
AK303195.1
-
6
577
AK315911.1
-
7
638
AK316298.1
-
7
682
AL080116.1
-
7
690
BC035494.2
-
7
666
4298339
BC047689.1
-
7
667
CR456814.1
-
2
184
U50950.1
-
3
317
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
nucleic acid metabolism protein
replication origin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
DNA replication
pre-replicative complex assembly involved in nuclear cell cycle DNA replication
DNA replication initiation
neural precursor cell proliferation
DNA replication origin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7cf5v:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0