Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130852.1 | NP_001124324.1 | 8, 9 | 1287 | ||
NM_012116.3 | NP_036248.3 | 9, 10 | 1425 | |||
GenBank mRNA | AB028645.1 | - | 9, 10 | 1403 | ||
AF117646.1 | - | 9, 10 | 1425 | |||
AF117647.1 | - | 8, 9 | 1287 | |||
BC028915.1 | - | 9, 10 | 1383 | 4777669 |