Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031696.3 | NP_001026866.1 | 4, 5 | 492 | ||
NM_001291311.1 | NP_001278240.1 | 4, 5 | 426 | |||
NM_012268.3 | NP_036400.2 | 4, 5 | 716 | |||
XM_005258704.1 | XP_005258761.1 | 4, 5 | 637 | |||
XM_005258707.4 | XP_005258764.1 | 4, 5 | 419 | |||
XM_005258708.3 | XP_005258765.1 | 3, 4 | 312 | |||
XM_005258709.4 | XP_005258766.1 | 3, 4 | 300 | |||
XM_005258710.4 | XP_005258767.1 | 3, 4 | 341 | |||
XM_006723122.1 | XP_006723185.1 | 4, 5 | 804 | |||
XM_011526692.1 | XP_011524994.1 | 4, 5 | 799 | |||
XM_011526693.1 | XP_011524995.1 | 4, 5 | 519 | |||
XM_017026546.1 | XP_016882035.1 | 5, 6 | 735 | |||
XM_017026547.1 | XP_016882036.1 | 4, 5 | 303 | |||
XM_017026548.1 | XP_016882037.1 | 4, 5 | 454 | |||
XM_017026549.1 | XP_016882038.1 | 4, 5 | 557 | |||
GenBank mRNA | AK293690.1 | - | 3, 4 | 217 | ||
BC036327.1 | - | 4, 5 | 217 | 5189261 | ||
BC096820.1 | - | 4, 5 | 415 | 30558682 | ||
BP304863.1 | - | 3, 4 | 232 | |||
U60644.1 | - | 4, 5 | 423 |